Check Order Status
Carbon Steel Sterile Scalpel Blade
Actual product may vary
Fisherbrand
Carbon Steel Sterile Scalpel Blade
Have Questions?
Catalog Number
Size
FKS11882431
# 25
Catalog number
FKS11882431
Price (SGD)
88.00
PK
Add to cart
Size:
# 25
Price (SGD)
88.00
PK
Add to cart
Specifications
Blade Material
Carbon Steel
Product Type
Scalpel Blade
Size
# 25
For Use With (Equipment)
Fits # 4, # 4L, # 4 Graduated and # 6B Handles
Description
Surgeon Grade
Sterility
Sterile
Quantity
10/Pk.