Procare Hand Cream
Procare Hand Cream
Actual product may vary
Fisherbrand
Procare Hand Cream
Destroy a broad specturm of organisms with a hand cream specially formulated for long-lasting skin care protection. Day Impex™ ProCARE Hand Cream is a non-greasy professional barrier hand cream that relieves and protects hands from chapping and irritation.
Have Questions?
Catalog number FKS11877462
Price (SGD)
461.00
PK
Add to cart
Price (SGD)
461.00
PK
Add to cart
  • Particularly effective in reducing the irritation caused by the constant wearing of rubber gloves
  • Combining lanolin to moisturise the skin and silicone to seal in that moisture
  • Absorbs rapidly for a non-greasy feel and instant protection.
  • Thick cream formulation so it can be used sparingly.
  • A single 85 g tube will last several weeks
  • In biohazardous situations, we recommend a dedicated tube for each user to reduce tbe risk of cross-contamination
Specifications
Product TypeProcare Hand Cream
ColorOff-white
FormCream
CompositionCetyl Alcohol Dimethicone, Sorbitol Glyceryl Stearate, Sodium Lauryl Sulphate PEG-60 Lanolin, Nonoxynol-9 (permissible minimum) Propylene Glycol, Diazolidinyl Urea, Methylparaben, Propylparaben Water, Kathon CG
For Use With (Application)Laboratory and Healthcare Personnel
PackagingPump Top Dispenser
Quantity4 Pk.
Volume (Metric)450 g
Unit SizePK