Cardboard Cryoboxes, 133mm
Cardboard Cryoboxes, 133mm
Actual product may vary
Fisher Catalogue
Cardboard Cryoboxes, 133mm
Combine these boxes with different grid inserts to hold a wide range of tube sizes. Fisherbrand™ Cardboard Cryoboxes are temperature resistant down to -80°C, ideal for ultra-low temperature storage.
Have Questions?
Catalog NumberColorHeight (Metric)Capacity (Metric)Dimensions (L x W x H)
FKS11872473White50 mm2 mL133 x 133 x 50 mm
Catalog number FKS11872473
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
White
Height (Metric):
50 mm
Capacity (Metric):
2 mL
Dimensions (L x W x H):
133 x 133 x 50 mm
  • Available with a water-repellent finish or with a waterproof plastic coating
  • Highest flexibility: Cryoboxes can be combined with different grid inserts for tubes from 6 to 28.5mm in diameter
  • The Cryoboxes can be used for larger vessels if the grid is removed
Specifications
Capacity (Metric)2 mL
ColorWhite
MaterialCardboard
ShapeSquare
TypeCryo Box
Holds1 to 2 mL
Quantity10 Pack
Dimensions (L x W)133 x 133 mm
Dimensions (L x W x H)133 x 133 x 50 mm
Length (Metric)133 mm
Width (Metric)133 mm
Width (Metric) Interior133 mm
Height (Metric)50 mm
Weight (Metric)0.74 kg
Unit SizePK