Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes
Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes
Actual product may vary
Fisherbrand
Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes
Measure, mix and pour using 5.1 Borosilicate Glass Disposable Test Tubes. Tubes are rimmed and are available in various lengths.
Have Questions?
Catalog NumberDiameter (Metric)
FKS1187235325 mm
Catalog number FKS11872353
Price (SGD)
60.00
PK
Add to cart
Diameter (Metric):
25 mm
Price (SGD)
60.00
PK
Add to cart
  • Disposable
  • Clear borosilicate glass
  • Round bottom
  • Rimmed
Specifications
Certifications/ComplianceDIN 12395
Design TypeRound Bottom
DescriptionTest Tube
Length (Metric)150 mm
MaterialBorosilicate Glass
Quantity50 Pack
RimmedYes
AutoclavableAutoclavable
Diameter (Metric)25 mm
Unit SizePK