Eutech™ Precision pH-millivolt Simulator, Hi-Low impedence
Product Image
Fisherbrand
Eutech™ Precision pH-millivolt Simulator, Hi-Low impedence
The Eutech pH-millivolt simulator is a handy, easy-to-use service tool for testing the accuracy of your pH meter.
Have Questions?
Catalog NumberRange (mV)
FKS11871253±2
Catalog number FKS11871253
Price (SGD)
-
Range (mV):
±2
The Eutech pH-millivolt Simulator is a handy, easy-to-use service tool for testing the accuracy of your pH meter. Used for trouble-shooting and fault diagnosis of meters, the simulator works with most pH/redox instruments with BNC connectors. Simulator comes with a BNC cable and a sturdy rubber boot which doubles up as a stand.
Features
  • Push button pH/mV selection
  • pH accuracy of ±0.05; mV range of ±2
  • USA/NIST pH buffer standards simulation
  • 16 combination values
  • 1 GΩ high input impedance test
  • Large display with bright LED
  • Splashproof keypad
  • Protective rubber boot doubles up as stand
  • Power-saving meter runs on 4 AAA x 1.4 V batteries
  • Specifications
    Accuracy (pH)±0.05
    Display TypeLED
    Measurement ModespH, Meter, Accuracy, Test, mV
    Range (mV)±2
    Power Supply4 - AAA x 1.4V batteries
    Unit SizeEA