Erlenmeyer Flasks with Conical Ground Joint
Erlenmeyer Flasks with Conical Ground Joint
Actual product may vary
Erlenmeyer Flasks with Conical Ground Joint
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Have Questions?
Catalog NumberCapacity (Metric)Socket Joint
FKS1187048210 mL14/23 NS
Catalog number FKS11870482
Price (SGD)
233.00
PK
Add to cart
Capacity (Metric):
10 mL
Socket Joint:
14/23 NS
Price (SGD)
233.00
PK
Add to cart
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Certifications/ComplianceDIN EN ISO 4797
DescriptionConical Ground Joint, Inscription Label
Capacity (Metric)10 mL
MaterialBorosilicate Glass, Duran Glass
Socket Joint14/23 NS
Diameter (Metric) Outer30 mm
Unit SizePK