Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Includes |
---|---|---|
FKS11868003 | 2 L | Carrying Handle, Vented Secure Lid |
2122 | 1 L | Carrying Handle, Vented Secure Lid |
2123 | 2 L | Carrying Handle, Vented Secure Lid |
FIS11-670-4B | 2 L | Carrying Handle, Vented Secure Lid |
FIS13-262-780 | 1 L | Without Lid and Handle |
FIS13-762-372 | 1.9 L | Without Lid and Handle |
Shallow Wide Mouth: