Rubber Conical Gasket Guko, from EPDM, conical rubber gaskets, for filtering flasks
Rubber Conical Gasket Guko, from EPDM, conical rubber gaskets, for filtering flasks
Actual product may vary
Fisherbrand
Rubber Conical Gasket Guko, from EPDM, conical rubber gaskets, for filtering flasks
The rubber sleeve is required to achieve gas tightness during vacuum filtration.
Have Questions?
Catalog NumberSize (Metric)
FKS11860990Top dia: 29 mm; Bottom dia:16 mm
Catalog number FKS11860990
Price (SGD)
160.00
PK
Add to cart
Size (Metric):
Top dia: 29 mm; Bottom dia:16 mm
Price (SGD)
160.00
PK
Add to cart
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
  • For tight closure in vacuum filtration applications
  • Conical rubber gaskets
  • for filtering flasks
Specifications
For Use With (Application)Sealing; Filtering
Height (Metric)23 mm
Quantity10/Pk.
Size (Metric)Top dia: 29 mm; Bottom dia:16 mm
Width (Metric)3.5 mm wall thickness
Diameter (Metric) Outside16 mm
MaterialEPDM
Unit SizePK