Check Order Status
Cardboard Cryobox Inserts, 133mm
Actual product may vary
Fisherbrand
Cardboard Cryobox Inserts, 133mm
Combine these inserts with Cryoboxes for flexible cell storage and sample handling. Fisherbrand™ Cardboard Cryobox Inserts feature ultra-low temperature resistant cardboard with water repellent finish for sample storage.
Have Questions?
Change view
Catalog Number
Height (Metric)
Array
FKS11852513
30mm
10 x 10
FKS11842513
25mm
10 x 10
FKS11802513
25mm
9 x 9
3 Options
Catalog number
FKS11852513
Price (SGD)
76.00
PK
Add to cart
Height (Metric):
30mm
Array:
10 x 10
Price (SGD)
76.00
PK
Add to cart
Ultra low temperature-resistant cardboard
For sample storage
Temperature-resistant down to -80°C
Water-repellent finish
Specifications
Quantity
10 Pack
Material
Cardboard
Product Type
Cryo Box Insert
Height (Metric)
30mm
Width (Metric)
133mm
Length (Metric)
133mm
Array
10 x 10