Check Order Status
Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes
Actual product may vary
Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes
Measure, mix and pour using 5.1 Borosilicate Glass Disposable Test Tubes. Tubes are rimmed and are available in various lengths.
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric)
FKS11852363
75 mm
10 mm
Catalog number
FKS11852363
Price (SGD)
57.00
PK
Add to cart
Length (Metric):
75 mm
Diameter (Metric):
10 mm
Price (SGD)
57.00
PK
Add to cart
Disposable
Clear borosilicate glass
Round bottom
Rimmed
Specifications
Design Type
Round bottom
Length (Metric)
75 mm
Packaging
100Pack
Rimmed
Yes
Diameter (Metric)
10 mm
Product Type
Test Tube