Class A PMP Measuring Cylinders
Class A PMP Measuring Cylinders
Actual product may vary
Fisherbrand
Class A PMP Measuring Cylinders
Manufactured from transparent polymethypentene (PMP) and complies with DIN 12681 and ISO 6706 standards. BRAND™ Class A PMP Measuring Cylinders include DE-M markings in a particularly high-quality printing ink. Exposure to temperatures up to 121°C (autoclaving) will not cause permanently exceeded error limits. Cleaning up to max. 60°C is recommended to preserve marks and inscriptions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS1184833150 mL1 mL
FKS107495311000 mL10 mL
Catalog number FKS11848331
Price (SGD)
84.00
PK
Add to cart
Capacity (Metric):
50 mL
Graduations:
1 mL
Price (SGD)
84.00
PK
Add to cart
  • Manufactured from transparent polymethypentene (PMP.)
  • Complies with DIN 12681 and ISO 6706 standards.
  • Calibrated to contain (TC, In).
  • Includes one batch certificate.
  • DE-M markings in a particularly high-quality printing ink used in the BRAND™ graduated cylinders.
  • Exposure to temperatures up to 121°C (autoclaving) will not cause permanently exceeded error limits.
  • Cleaning up to max. 60°C is recommended to preserve marks and inscriptions.
Specifications
Capacity (Metric)50 mL
MaterialPMP
Tolerance±0.5 mL
Item DescriptionDE-M Markings in high-quality blue ink
Certifications/ComplianceDIN 12681, ISO 6706
No. per Pack1Pack
ClassA
CalibrationTo Contain
IncludesBatch Certificate
Quantity1Pack
Graduations1 mL
AutoclavableAutoclavable
Limit of Error±0.5mL
Max. Temperature121°C
Subdivision1mL
ShapeRound
Scale ColorBlue
Base TypeHexagonal
Height (Metric)200 mm
TypeGraduated Measuring Cylinder
Unit SizePK