Check Order Status
18/10 Stainless Steel Dissecting Needle
Actual product may vary
Fisherbrand
18/10 Stainless Steel Dissecting Needle
For use with Bochem needle holder Kolle 12000. Bochem™ 18/10 Stainless Steel Dissecting Needle is manufactured from stainless steel.
Have Questions?
Catalog Number
For Use With (Equipment)
Diameter (Metric) Inside
FKS11845101
Type 1 Eye Holder KOLLE 12000
1.5 mm
Catalog number
FKS11845101
Price (SGD)
77.00
PK
Add to cart
For Use With (Equipment):
Type 1 Eye Holder KOLLE 12000
Diameter (Metric) Inside:
1.5 mm
Price (SGD)
77.00
PK
Add to cart
Type 1: eye
Type 2: needle
Type 3: lancet
For use with Bochem needle holder Kolle 12000
Manufactured from stainless steel
Specifications
Diameter (Metric) Inside
1.5 mm
Length (Metric)
100 mm
Material
Stainless Steel (18/10)
Product Type
Eye
For Use With (Equipment)
Type 1 Eye Holder KOLLE 12000
Quantity
10/pk.