Check Order Status
Frosted Margin Soda Lime Glass Slide
Actual product may vary
Fisherbrand
Frosted Margin Soda Lime Glass Slide
Holds specimens for observation with a microscope. Fisherbrand™ Frosted Glass Microscope Slides are manufactured from soda lime glass with a frosted margin.
Have Questions?
Catalog Number
Thickness (Metric)
FKS11844782
1.1 mm
Catalog number
FKS11844782
Price (SGD)
25.00
PK
Add to cart
Thickness (Metric):
1.1 mm
Price (SGD)
25.00
PK
Add to cart
Dimensions: approx. 76 x 26mm
Supplied in packs of 50
Specifications
Edges
90° or Raw Lapped
Frosted
Margin
Length (Metric)
76 mm
Material
Soda Lime Glass
Thickness (Metric)
1.1 mm
Width (Metric)
26 mm
Coating
Polished
Quantity
50 Pack