Fireboy™ Eco Bunsen Burner
Fireboy™ Eco Bunsen Burner
Actual product may vary
Fisherbrand
Fireboy™ Eco Bunsen Burner
Ensures highest application safety with continuous flame monitoring, overtemperature protection and user-defined, maximal burning times. Fireboy™ Eco Bunsen Burner features smooth, chrome-plated metal housing which is easy to clean and both UV- and solvent-resistant.
Have Questions?
Catalog NumberGas Type
FKS11843560Natural Gas, Butane, Propane
Catalog number FKS11843560
Price (SGD)
3,058.00
EA
Add to cart
Gas Type:
Natural Gas, Butane, Propane
Price (SGD)
3,058.00
EA
Add to cart
The smooth, chrome plated metal housing is easy to clean and both UV and solvent resistant. With flame monitor and overheating protection. Functional and robust design. Convenient handling. Safe operation.
  • Ignition by a footswitch (included) or push button
  • Two adjustment knobs for air and gas allow easy fine-tuning of flame FIREBOY plus
  • Ignition by push button, or hands free operation with movement sensor or optional footswitch (not included)
  • User defined maximal burning times
  • Optional rechargeable battery
  • Portable Compatible with common gas types such as butane/propane and natural gas; can be connected to gas distribution system or gas cartridges.
Specifications
Product TypeBunsen Burner
Gas TypeNatural Gas, Butane, Propane
Description100W x 150D mm
Diameter (Metric)150 mm
IncludesFoot Switch, (2) Nozzle, Adapter
No. per Pack1 Ea.
Width (Metric)100 mm
Unit SizeEA