Check Order Status
Foam Stands for Vortex Mixer
Actual product may vary
Fisherbrand
Foam Stands for Vortex Mixer
Securely holds test tubes and vials. Velp Scientifica™ Foam Stands are compatible with Velp Scientifica Vortex Mixers.
Have Questions?
Catalog Number
Holds
No. of Positions
FKS11840992
29 mm Tubes
4
Catalog number
FKS11840992
Price (SGD)
232.00
EA
Add to cart
Holds:
29 mm Tubes
No. of Positions:
4
Price (SGD)
232.00
EA
Add to cart
Black in color
Soft foam to prevent breakage
Available in different diameters and capacities
Specifications
Holds
29 mm Tubes
Tube Size
29 mm
Product Type
Foam Tube Rack
For Use With (Equipment)
CLASSIC, ZX3, WIZARD, ZX4, TX4
No. of Positions
4