Polypropylene Culture Tubes
Polypropylene Culture Tubes
Actual product may vary
Fisherbrand
Polypropylene Culture Tubes
Non-sterile Culture Tubes without Caps
Have Questions?
Catalog NumberCapacity (Metric)
FKS1183864116 mL
Catalog number FKS11838641
Price (SGD)
-
Capacity (Metric):
16 mL
  • Natural polypropylene
  • Non-sterile, without cap
  • 17mm x 100mm (Dia., x H)
Specifications
Diameter (Metric)17 mm
Closure Size17 mm dia.
DescriptionHeavy walled, break resistant culture tubes without caps
ColorNatural
Length (Metric)100 mm
MaterialPolypropylene
SterilityNon-sterile
Capacity (Metric)16 mL
Closure IncludedNo
Unit SizePK