Inoculating L-Shaped Spreaders
Inoculating L-Shaped Spreaders
Actual product may vary
Fisherbrand
Inoculating L-Shaped Spreaders
For spreading over the surface of solid media in microbiological applications. Gosselin™ Inoculating L-Shaped Spreaders are manufactured from polystyrene.
Have Questions?
Catalog NumberQuantity
FKS11836191Subpacks of 5
Catalog number FKS11836191
Price (SGD)
147.00
PK
Add to cart
Quantity:
Subpacks of 5
Price (SGD)
147.00
PK
Add to cart
  • Curved end
  • Sterile and disposable
  • Packed in tamper-evident ZIP bags
Specifications
Certifications/ComplianceEC N°1935/2004, ISO 11137
ColorWhite
DisposableSingle-use
Length (Metric)148 mm
QuantitySubpacks of 5
ShapeL-Shape
MaterialPolystyrene
Product TypeInoculating Spreader
SterilitySterile
For Use With (Application)For Dairy, Environmental, Industrial, Microbiological Uses
Unit SizePK