Autoclavable Bags
Autoclavable Bags
Actual product may vary
Fisher Catalogue
Autoclavable Bags
For autoclaving contaminated products safely
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)MaterialWidth (Metric)Height (Metric)
FKS118304823 LPolypropylene25.5 cm40 cm
FIS07-201-9639 LPolypropylene30 cm50 cm
FIS07-201-96514 LPolypropylene40 cm60 cm
Catalog number FKS11830482
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
3 L
Material:
Polypropylene
Width (Metric):
25.5 cm
Height (Metric):
40 cm
  • Polypropylene (PP) resistant bags
  • Thermal resistance 134°C
  • Transparent
Specifications
Capacity (Metric)3 L
Capacity (English)0.79 gal.
ColorNatural
Height (Metric)40 cm
Quantity500/Pk.
Width (Metric)25.5 cm
AutoclavableAutoclavable
For Use With (Application)Clinical, dairy, environment, industries, microbiology
Width (English)10.03 in.
Thickness40 μm
Height (English)15.74 in.