Check Order Status
Pyrex™ Borosilicate Graduated Glass dropping Funnels with Cone and PTFE Key
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Graduated Glass dropping Funnels with Cone and PTFE Key
Has double graduated scale showing volume of liquid contained/delivered
Have Questions?
Catalog Number
Joint Number
Graduations
FKS11830341
29/32
10mL
Catalog number
FKS11830341
Price (SGD)
537.00
PK
Add to cart
Joint Number:
29/32
Graduations:
10mL
Price (SGD)
537.00
PK
Add to cart
Has double graduated scale showing volume of liquid contained/delivered
Complies with ISO 4800
Has interchangeable PTFE key for grease free performance
Stem inside cone for easy fitting to Quickfit™ assemblies
Graduations in white enamel
Specifications
Capacity (Metric)
500 mL
Material
Borosilicate Glass
Certifications/Compliance
ISO 4800
Description
PTFE closure
Graduated
White enamel
Includes
Cone
Graduations
10mL
Joint Number
29/32
Product Type
Dropping Funnel