Sterile Disposable Standard Serological Pipets
Sterile Disposable Standard Serological Pipets
Actual product may vary
Sterile Disposable Standard Serological Pipets
Sterile Disposable Standard Serological Pipets
Sterile Disposable Standard Serological Pipets
Fisherbrand
Sterile Disposable Standard Serological Pipets
Polystyrene pipet with negative graduations on all sizes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)GraduationsTip StyleTolerance
FKS1182967050 mL0.5 mLStandard±1 mL
FIS13-678-12B1 mL0.01 mLStandard±0.02 mL
FIS13-678-12C2 mL0.01 mLStandard±0.04 mL
FIS13-678-12D5 mL0.10 mLStandard±0.1 mL
FIS13-678-12E10 mL0.1 mLStandard±0.2 mL
FKS1180967010 mL0.1 mLStandard±0.2 mL
FIS13-678-14B25 mL0.2 mLStandard±0.5 mL
FIS13-678-14C50 mL0.5 mLStandard±1 mL
Catalog number FKS11829670
Price (SGD)
425.00
PK
Add to cart
Volume (Metric):
50 mL
Graduations:
0.5 mL
Tip Style:
Standard
Tolerance:
±1 mL
Price (SGD)
425.00
PK
Add to cart
Fisherbrand™ Sterile Disposable Standard Serological Pipets feature a standard tip. Polystyrene construction, with negative graduations on all sizes. Certified ASTM E934, ASTM E1380 compliant.
  • Ascending and descending graduations on all but the 1 and 2mL sizes
  • Sterile and nonwettable pipets
  • Offer a tolerance of ±2% at full volume and have plug-in mouth end
  • Without magnifier stripe
  • Recyclable outer packaging
  • Bulk packed
  • Each pipet lot is tested for sterility, accuracy and endotoxin level
Specifications
Certifications/ComplianceASTM E934, ASTM E1380; DNase/RNase Free; BSE/TSE Free
MaterialPolystyrene
SterilitySterile
GraduatedYes
Graduations0.5 mL
Negative Graduations9 mL
Quantity100/Cs., 25/Pk., 4 Packs/Cs.
Tip StyleStandard
Tolerance±1 mL
Packaging TypeBulk
Volume (English)1.69 oz.
ColorPurple
Volume (Metric)50 mL
TypeSerological Pipet