BLAUBRAND™ AR-GLAS™ Type 3 Class AS Measuring Pipets
BLAUBRAND™ AR-GLAS™ Type 3 Class AS Measuring Pipets
Actual product may vary
Fisherbrand
BLAUBRAND™ AR-GLAS™ Type 3 Class AS Measuring Pipets
Manufactured from AR-GLAS™ Soda lime glass. BRAND™ Blaubrand™ AR-GLAS™ Type 3 Class AS Measuring Pipets include graduations with DE-M marking and are calibrated for total delivery. Type 3 with zero on top.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)GraduationsTolerance
FKS1182831150 mL0.5 mL±0.20 mL
FKS1180830125 mL0.1 mL±0.100 mL
Catalog number FKS11828311
Price (SGD)
437.00
PK
Add to cart
Volume (Metric):
50 mL
Graduations:
0.5 mL
Tolerance:
±0.20 mL
Price (SGD)
437.00
PK
Add to cart
  • Manufactured from AR-GLAS™ Soda lime glass.
  • Type 3 (zero on top)
  • DE-M Marking graduations
  • Calibrated to deliver (TD, Ex.)
  • Supplied with batch number and one batch certificate per packing unit
  • USP individual certificate or DAkkS calibration certificate available upon request.
Specifications
Graduations0.5 mL
IncludesCotton Plug
Tolerance±0.20 mL
Volume (Metric)50 mL
Unit SizePK