Polypropylene Cylindrical T Connectors
Polypropylene Cylindrical T Connectors
Actual product may vary
Fisherbrand
Polypropylene Cylindrical T Connectors
Manufactured from polypropylene for high thermal resistance. Buerkle™ Polypropylene Cylindrical T Connectors resist temperatures up to 135°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
FKS118265837 to 9 mm Hose
FKS1005495011 to 13 mm Hose
Catalog number FKS11826583
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With:
7 to 9 mm Hose
Cylindrical cross angle hose connector
  • Polypropylene
  • Autoclavable to 121°C
  • Suitable for foodstuff
  • Available in a wide range of diameters for varying hose sizes
Specifications
Certifications/ComplianceEU Foodstuffs, RoHS
AutoclavableAutoclavable
MaterialPolypropylene
For Use With7 to 9 mm Hose
Packaging Quantity10/Pk.; 20/Scale
Resistance PropertiesUp to 135°C Temperature-Resistant
ShapeCylindrical T
ColorTransparent
Max. Operating Temperature (Metric)135°C
Product TypeT-Connector
Unit SizePK