Elite™ Variable Volume, Multichannel Pipettes
Elite™ Variable Volume, Multichannel Pipettes
Actual product may vary
Fisherbrand
Elite™ Variable Volume, Multichannel Pipettes
New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)AccuracyNumber of Channels
FKS1182577210 to 100 μL1.3 to 5%8
FISFBE8000505 to 50 μL1.5 to 5%8
FISFBE80030030 to 300 μL1 to 4.6%8
Catalog number FKS11825772
Price (SGD)
1,960.00
EA
Add to cart
Volume (Metric):
10 to 100 μL
Accuracy:
1.3 to 5%
Number of Channels:
8
Price (SGD)
1,960.00
EA
Add to cart
New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort
  • Fully autoclavable - entire pipetter can be steam autoclaved
  • Extremely low plunger forces - work longer without fatigue and reduce the risk of RSI
  • Soft-touch tip ejection—a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle - comfortable grip and effortless tip attachment
  • Optimal fit with SureOne pipetter tips - best pipetting performance
  • Super blow-out piston - creates 150% bigger air push than conventional piston for delivery of the micro size drops
Specifications
Accuracy1.3 to 5%
Compatible TipsSureOne™
AutoclavableAutoclavable
For Use WithSureOne Pipette Tip
Number of Channels8
Product LineElite
TypeVariable Volume Pipette
Volume (Metric)10 to 100 μL
Finger StyleErgonomic
Increments0.2μL
Unit SizeEA