Nickel Chattaway Medium Spatulas
Nickel Chattaway Medium  Spatulas
Actual product may vary
Fisherbrand
Nickel Chattaway Medium Spatulas
Bright finish, blade at one end flat, and a 45 degree angle at the other end; these spatulas are ideal for use with a wide range of compounds in most laboratory environments.
Have Questions?
Catalog NumberLength (Metric)
FKS11824593180 mm
Catalog number FKS11824593
Price (SGD)
204.00
PK
Add to cart
Length (Metric):
180 mm
Price (SGD)
204.00
PK
Add to cart
  • Nickel 200 spec (minimum 99.6% nickel content)
  • Ideal for use with a wide range of compound.Bright finish
  • Blade at one end flat, other end at 45° angle 
  • 'Micro weight' - centre barrel diameter 1.60mm
  • 'Medium weight' - centre barrel diameter 3.65mm
  • Long enough to protect user's hands from hot surfaces
Specifications
Quantity5 Pk.
Product TypeSpatula
FinishBright
For Use With (Application)For Structural Applications Where Resistance to Corrosion is Prime Consideration
Width (Metric) Blade10 mm
Temperature (Metric) Max315°C
MaterialNickel
Length (Metric)180 mm
Unit SizePK