Blaubrand™ Eterna™ Class AS Bulb Pipets
Blaubrand™ Eterna™ Class AS Bulb Pipets
Actual product may vary
Fisherbrand
Blaubrand™ Eterna™ Class AS Bulb Pipets
Manufactured from AR-GLAS™ Soda-lime glass and complies with DIN EN ISO 648 standards. BRAND™ Blaubrand™ Eterna™ Class AS Bulb Pipets are delivered with batch number and one batch certificate per packing unit of the manufacturer. USP Individual certificate or DAkkS calibration certificate available by request.
Have Questions?
Catalog NumberVolume (Metric)ToleranceColor
FKS118239801 mL0.008 mLBlue
Catalog number FKS11823980
Price (SGD)
250.00
PK
Add to cart
Volume (Metric):
1 mL
Tolerance:
0.008 mL
Color:
Blue
Price (SGD)
250.00
PK
Add to cart
  • Manufactured from AR-GLAS™ Soda-lime glass.
  • DIN EN ISO 648. Calibrated to deliver (TD, Ex).
  • Waiting time 5 seconds.
  • Delivered with batch number and one batch certificate per packing unit of the manufacturer.
  • On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
Specifications
CalibrationTo Deliver
Certifications/ComplianceDIN EN ISO 648, DIN 12691
ClassAS
MaterialAR-GLAS, Soda Lime Glass
GraduatedYes
Graduation ColorAmber
Graduation TypeDE-M Marking
Quantity12 Pack
Tolerance0.008 mL
ColorBlue
Volume (Metric)1 mL
TypeVolumetric Bulb Pipet
Unit SizePK