Acrylic Glove Box Holder
Acrylic Glove Box Holder
Actual product may vary
Fisherbrand
Acrylic Glove Box Holder
Keep gloves organized and ready for immediate use. Fisherbrand™ Acrylic Glove Box Holder includes a foam insert ensures a secure fit and easy dispensing.
Have Questions?
Catalog NumberCapacityDimensions (D x W x H) Interior
FKS118171123 boxes110 x 295 x 385 mm
Catalog number FKS11817112
Price (SGD)
410.00
EA
Add to cart
Capacity:
3 boxes
Dimensions (D x W x H) Interior:
110 x 295 x 385 mm
Price (SGD)
410.00
EA
Add to cart
  • Clear acrylic holders include a polyester foam insert to securely hold a range of glove box sizes–snug fit ensures easy removal of gloves
  • All holders fit most major brands of boxes
  • Mounting screws included
Specifications
Product TypeGlove Box Holder
ColorClear
Depth (Metric)110 mm
Dimensions (D x W x H) Interior110 x 295 x 385 mm
Width (Metric)295 mm
Height (Metric)260 mm
MaterialAcrylic
Capacity3 boxes
IncludesWall Mount Screw
Quantity1 Ea.
Unit SizeEA