BioLite™ Cell Culture Treated Dishes
BioLite™ Cell Culture Treated Dishes
Actual product may vary
Fisherbrand
BioLite™ Cell Culture Treated Dishes
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite™ Cell Culture Treated Dishes that are guaranteed to be sterile, non-pyrogenic, and non-cytotoxic.
Have Questions?
Catalog NumberDiameter (Metric)
FKS11815275100 mm
Catalog number FKS11815275
Price (SGD)
255.00
PK
Add to cart
Diameter (Metric):
100 mm
Price (SGD)
255.00
PK
Add to cart
Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated dishes suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.
• Cell culture treated
• Sterile per ISO 11137, SAL 10-6
• USP Class VI and non-cytotoxic
• Non-pyrogenic
Specifications
Bottom ShapeFlat
Capacity (Metric)12.5 mL (Well)
Culture Area60.8 sq. cm
MaterialPolyethylene
ShapeRound
SterilitySterile
Surface TreatmentCell Culture
TypeCell Culture Dish
Volume (Metric) Working12.5 mL
No. per Case200
No. per Pack10
Height (Metric)20 mm
Height (Metric) Interior16 mm
Diameter (Metric)100 mm
Unit SizePK