Wide Mouth Containers with Handle
Wide Mouth Containers with Handle
Actual product may vary
Fisherbrand
Wide Mouth Containers with Handle
Container with 2.3 l and 4.4 l contents and particularly wide opening. Therefore ideal for filling with bulk goods of all kinds. Products such as spices, tea, granulates can be packaged, stored or displayed as samples in these containers. Space-saving square shape.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS118136824400 mL
FKS118036822300 mL
Catalog number FKS11813682
Price (SGD)
80.00
EA
Add to cart
Capacity (Metric):
4400 mL
Price (SGD)
80.00
EA
Add to cart
  • Safe handling with integral handle
  • Transparent polyethylene provides good view of contents.
  • Leakproof wide-necked lid
  • 0317-2300 with tamper-evident screw cap made of blue PP, 0317-4400 with screw cap made of LDPE
Specifications
Capacity (Metric)4400 mL
ColorTransparent
Diameter (Metric) Inner Neck109 mm
IncludesLDPE screw cap
MaterialPolyethylene
Length (Metric)142 mm
Quantity1
Width (Metric)142 mm
Thread Size (Metric)120 mm
Closure IncludedYes
MouthWide
Height (Metric)263 mm
Closure MaterialLDPE
Closure TypeScrew Cap
Unit SizeEA