Captair™ Replacement Filters
Captair™ Replacement Filters
Actual product may vary
Captair™ Replacement Filters
Captair™ Replacement Filters
Fisherbrand
Captair™ Replacement Filters
A range of replacement filters for use with fume hoods and vented cabinets.
Have Questions?
Catalog NumberFor Use With (Equipment)For Use With (Application)
FKS11812934Fume HoodsFor Organic Vapors
Catalog number FKS11812934
Price (SGD)
1,533.00
EA
Add to cart
For Use With (Equipment):
Fume Hoods
For Use With (Application):
For Organic Vapors
Price (SGD)
1,533.00
EA
Add to cart
  • For a wide variety of fume hoods and cabinets
  • Filters for organic vapors, acid vapors and more
Specifications
For Use With (Equipment)Fume Hoods
Product TypeFilter
For Use With (Application)For Organic Vapors
Unit SizeEA