Check Order Status
Kimble Chase™ Borosilicate Glass Test Tubes, Disposable
Actual product may vary
Kimble Chase™ Borosilicate Glass Test Tubes, Disposable
Thick-walled for particularly good mechanical resistance.
Have Questions?
Catalog Number
Diameter (Metric)
FKS11812373
16 mm
Catalog number
FKS11812373
Price (SGD)
72.00
PK
Add to cart
Diameter (Metric):
16 mm
Price (SGD)
72.00
PK
Add to cart
DIN 12395
Excellent resistance to thermal shock
Autoclavable
Glass type I complies with USP/EP
Specifications
Description
Wall thickness of 0.5 to 0.6mm
Length (Metric)
160 mm
Material
Borosilicate Glass
Packaging
100Pack
Rimmed
Yes
Autoclavable
Autoclavable
Diameter (Metric)
16 mm
Product Type
Test Tube