Wire Centrifuge Bottle Rack
Wire Centrifuge Bottle Rack
Actual product may vary
Fisherbrand
Wire Centrifuge Bottle Rack
Accommodates large samples and bottles up to 60mm in diameter. Fisherbrand™ Wire Centrifuge Bottle Rack can be used with conical, round or flat-bottom vessels.
Have Questions?
Catalog NumberArray
FKS118058702 x 3
Catalog number FKS11805870
Price (SGD)
126.00
PK
Add to cart
Array:
2 x 3
Price (SGD)
126.00
PK
Add to cart
  • Holds bottles and vessels up to 60 mm dia
  • Chemically-resistant epoxy-coated steel wire racks
  • Suitable for conical, round, or flat bottom glass or plastic tubes and bottles
  • Autoclavable
  • Slotted bottom
Specifications
Product TypeRack
Array2 x 3
AutoclavableAutoclavable
MaterialEpoxy-coated Steel
For Use With (Application)Transport or sample handling
For Use With (Equipment)Vessels up to 60mm dia.
Quantity1 Each
Dimensions (L x W x H)205 x 135 x 104 mm
Length (Metric)205 mm
Width (Metric)135 mm
Height (Metric)104 mm
Diameter (Metric) Well60 mm
Unit SizePK