Cleargreen™ Non-DEHP PVC Tubing
Cleargreen™ Non-DEHP PVC Tubing
Actual product may vary
Cleargreen™ Non-DEHP PVC Tubing
Cleargreen™ Non-DEHP PVC Tubing
Fisherbrand
Cleargreen™ Non-DEHP PVC Tubing
Eliminate odor transferring to samples with this PVC tubing. Cleargreen™ Non-DEHP PVC Tubing is ideal for laboratory, food & beverage, pharmaceutical, environmental and general lab applications.
Have Questions?
Catalog NumberDiameter (Metric) Inner
FKS117996296.3 mm
Catalog number FKS11799629
Price (SGD)
416.00
EA
Add to cart
Diameter (Metric) Inner:
6.3 mm
Price (SGD)
416.00
EA
Add to cart
  • Odorless, phthalate free and translucent.
  • Low water absorption, low extractables
  • Certifications: FDA approved, USP Class VI, 3A compliant, REACH-compliant
  • Temperature range: -50 to 74°C
  • Sterilize: Autoclave or ethylene oxide
Specifications
Certifications/ComplianceFDA Compliant, USP Class VI, 3A compliant, Reach compliant and RoHS compliant
AutoclavableAutoclavable
Diameter (Metric) Inner6.3 mm
MaterialPVC
Diameter (Metric) Outer9.5 mm
Overall Length (Metric)15.24 m
Thickness (Metric)1.6 mm
For Use With (Application)Lab research, food and beverage, pharmaceutical, environmental and general lab applications
Min. Operating Temperature (Metric)-50°C
Packaging Quantity1 Each
SterilitySterile
ColorTranslucent
Max. Operating Temperature (Metric)74°C
Unit SizeEA