Sterilin™ Tubes with Screw Cap
Sterilin™ Tubes with Screw Cap
Actual product may vary
Fisherbrand
Sterilin™ Tubes with Screw Cap
Hold samples ranging from 5 to 10mL in these tubes with screw caps. Available in chemically resistant polypropylene or virgin polystyrene with a polyethylene screw cap.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Label TypeMaterialNo. per Case
FKS1179897810 mLPrinted LabelPolystyrene500
FKS117388685 mLNo LabelPolystyrene1000
Catalog number FKS11798978
Price (SGD)
1,618.00
PK
Add to cart
Capacity (Metric):
10 mL
Label Type:
Printed Label
Material:
Polystyrene
No. per Case:
500
Price (SGD)
1,618.00
PK
Add to cart
Hold samples ranging from 5 to 10mL in Thermo Scientific™ Sterilin™ Tubes with Screw Cap. Available in chemically resistant polypropylene or virgin polystyrene with a polyethylene screw cap. Supplied nonsterile.
Not available in all geographic regions.
Features:
  • Available labeled for sample identification
  • Supplied nonsterile
Manufactured from either:
  • Virgin polystyrene with flat base
  • Chemically resistant polypropylene with a skirted base
Ordering Information:
Product not available in all geographic regions.
Specifications
Product TypeSample Tube
Capacity (Metric)10 mL
MaterialPolystyrene
Label TypePrinted Label
No. per Case500
Product LineSterilin™
SterilityNon-sterile
Packaging Quantity500 Case
Closure MaterialPolyethylene
Capacity (English)0.3381 fl oz.
Closure TypeScrew Cap
PrintedYes
Unit SizePK