Petri Dishes
Petri Dishes
Actual product may vary
Petri Dishes
Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
Have Questions?
Catalog NumberDimensions (Dia. x H)Well CountVentSterility
FKS11798563120.5 x 15.8 mm14Sterile or Aseptic
Catalog number FKS11798563
Price (SGD)
570.00
PK
Add to cart
Dimensions (Dia. x H):
120.5 x 15.8 mm
Well Count:
1
Vent:
4
Sterility:
Sterile or Aseptic
Price (SGD)
570.00
PK
Add to cart
Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
  • Crystal polystyrene
  • Optimal flatness
  • Manufactured in class 100
  • Easy opening
Specifications
ColorTransparent
MaterialPolystyrene
ShapeSquare
SterilitySterile or Aseptic
Well Count1
PackagingRegular
Vent4
Dimensions (Dia. x H)120.5 x 15.8 mm
Height (Metric)15.8 mm
Diameter (Metric)120.5 mm
Unit SizePK