Vinyl Coated Lead Ring
Vinyl Coated Lead Ring
Actual product may vary
Fisherbrand
Vinyl Coated Lead Ring
Stabilize vessels and labware without scratching or marking them. Vinyl Coated Lead Ring features a soft coating around O-shaped lead rings. Available in four sizes to accommodate a variety of flask sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWeight (Metric)Diameter (Metric) InnerDiameter (Metric) OuterHolds
FKS11797884508 g51 mm90 mmFlasks (250 to 1000 mL)
FKS10195573277 g48 mm77 mmFlasks (125 to 500 mL)
FKS11707894635 g57 mm98 mmFlasks (500 to 2000 mL)
FKS11717894970 g70 mm115 mmFlasks (1 to 4 L)
Catalog number FKS11797884
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Weight (Metric):
508 g
Diameter (Metric) Inner:
51 mm
Diameter (Metric) Outer:
90 mm
Holds:
Flasks (250 to 1000 mL)
  • O-shaped rings stabilize vessels and labware.
  • Can also be used to immerse vessels in water baths.
  • Vinyl-coated lead rings will not mark or scratch benchtops and labware.
  • Available in a variety of sizes to fit a variety of flasks.
Specifications
Product TypeLead Ring
AutoclavableNot Autoclavable
ColorBlue
HoldsFlasks (250 to 1000 mL)
MaterialLead
ShapeCircular
ShelvesNone
SterilityNon-sterile
Quantity1 Ea.
Diameter (English) Inner2 in.
Diameter (English) Outer3.5 in.
Diameter (Metric) Outer90 mm
Diameter (Metric) Inner51 mm
Weight (English)17.9 oz.
Weight (Metric)508 g
Unit SizeEA