Vinyl Printer Labels
Vinyl Printer Labels
Actual product may vary
Fisherbrand
Vinyl Printer Labels
For use with label printer.
Have Questions?
Catalog NumberColorWidth (Metric)Height (Metric)Adhesive Type
FKS11797294White/Transparent25.4 mm19 mmAcrylic, Permanent
Catalog number FKS11797294
Price (SGD)
171.00
BOX
Add to cart
Color:
White/Transparent
Width (Metric):
25.4 mm
Height (Metric):
19 mm
Adhesive Type:
Acrylic, Permanent
Price (SGD)
171.00
BOX
Add to cart
  • Repositionable rubber-based adhesive
  • Semi-gloss finish
  • For panel identification and wire and cable marking
Specifications
Certifications/ComplianceRoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized
DescriptionBMP71 Label Printer Labels
Adhesive TypeAcrylic, Permanent
ColorWhite/Transparent
FinishMatte
FormatSelf-laminating
MaterialVinyl
Print TechnologyThermal Transfer
Product TypePrinter Label
For Use With (Application)Wire and cable identification
For Use With (Equipment)BMP™71
Labels per Row1
Quantity1000/Pk.
ShapeRectangle
Size (Metric)25.4W x 19 mmH
Temperature Range (Metric)-40°C to +70°C
Thickness (Metric)0.025 mm
Vertical Repeat (Metric)22.23 mm
Wire Gauge10 to 4 AWG
Dimensions (LxW)19.05 x 25.40 mm
Width (Metric)25.4 mm
Height (Metric)19 mm
Height (Metric) Printable Area9.53 mm
Unit SizeBOX