DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated
DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated
Actual product may vary
Fisherbrand
DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated
Glass laboratory bottles with superior thermal and chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11796878250 mL
FKS11706888500 mL
FKS117168881000 mL
Catalog number FKS11796878
Price (SGD)
1,112.00
PK
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
1,112.00
PK
Add to cart
The DURAN™ premium GL 45 laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. The premium bottles are ideal for high temperature applications: the thermal shock resistance of the glass bottle is certified for a 160°C differential, and is combined with a high temperature fluoropolymer (PFA) screw cap and pouring ring that are suitable for use up to 200°C. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature a double graduated scale for filling volume and a reversed scale for pouring. The marking field and Retrace Code for manufacturing lot traceability are printed in permanent white enamel.
  • Additional graduation marks and an opposite scaling (accurate to ± 5 %) simplify the reading of the filling level as well as the quantity withdrawn
  • Colourant free product
  • Reliable sealing due to PTFE-coated cap liner
  • TÜV tested thermal shock resistance of 160 K
  • Complete with PFA pouring ring and cap
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
Specifications
Capacity (Metric)250 mL
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage and transport of media and reagents; Shipping
IncludesWith premium screw cap and pouring ring
MaterialBorosilicate Glass (3.3)
Quantity10/Pk.
Thread Size (Metric)GL 45
AutoclavableAutoclavable
Closure MaterialPFA/PFA, PTFE/Silicone Layer
Diameter (Metric)70 mm
Unit SizePK