Ensure complete sample transfer to volumetric flasks with these reusable borosilicate glass weighing funnels. Cole-Parmer™ Glass Weighing Micro Funnels are flat on one side so you can set them on a balance.
Have Questions?
Catalog Number
Material
FKS11795308
Borosilicate Glass
Catalog number FKS11795308
Price (SGD)
586.00
PK
Add to cart
Material:
Borosilicate Glass
Price (SGD)
586.00
PK
Add to cart
Micro funnel is sized to fit microwell plates.
Specifications
MaterialBorosilicate Glass
For Use With (Application)Sample Transfer to Volumetric Flasks