Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Neck Finish |
---|---|---|
FKS11795253 | 250 mL | 43mm |
FKS11775243 | 30 mL | 28mm |
FIS02-896-2C | 125 mL | 38mm |
FIS02-896-2D | 250 mL | 43mm |
FIS02-896-2E | 500 mL | 53mm |
FIS02-896-2F | 1000 mL | 63mm |
FKS11528062 | 1000 mL | 63mm |
FKS11745253 | 1000 mL | 63mm |