1x1 image pixel for data collection
Anti-Static Static-Away™ Handle Brushes
Anti-Static Static-Away™ Handle Brushes
Actual product may vary
Fisherbrand
Anti-Static Static-Away™ Handle Brushes
Anti-Static Static-Away™ Handle Brushes designed with the softest hairs available to prevent scratching.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric) BrushLength (Metric) Handle
FKS117943465.08 cm12.7 cm
CTL15-078-2122.54 cm12.7 cm
CTL15-078-2135.08 cm12.7 cm
CTL15-078-2147.62 cm15.24 cm
FKS117043567.62 cm15.24 cm
Catalog number FKS11794346
Price (SGD)
111.00
EA
Add to cart
Length (Metric) Brush:
5.08 cm
Length (Metric) Handle:
12.7 cm
Price (SGD)
111.00
EA
Add to cart
Anti-Static Static-Away™ Handle Brushes designed with the softest hairs available to prevent scratching.
  • Remove static charges as they brush
  • Conductive composite handle
  • Long handles allow access to difficult places
  • Eliminate the possibility of a hand touching
  • Weight: <1 oz. (28.3 g)
Specifications
Product TypeAnti-static Brush
Bristle MaterialSynthetic Fibers
Bristle TypeSoft
Color-codedNo
Handle MaterialConductive Composite
Handle TypeAnti-static
Tip TypeStraight
ScentUnscented
SterilityNon-sterile
For Use With (Application)Ideal for dusting balances, cuvets, microscope slides, optics and instrument/computer screens, designed with the softest hairs available to prevent scratching
Length (English) Brush2 in.
Length (English) Handle5 in.
Length (Metric) Brush5.08 cm
Length (Metric) Handle12.7 cm
Weight (English)0.5 oz.
Weight (Metric)14.17 g
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link