Dust'It™ Blower
Dust'It™ Blower
Actual product may vary
Fisherbrand
Dust'It™ Blower
Powerful bulb duster forcibly blows away dust, lint and dirt with high-pressure, pinpoint jets of air.
Have Questions?
Catalog NumberDimensions (L x W)
FKS117943365.7 x 12.7 cm
Catalog number FKS11794336
Price (SGD)
-
Dimensions (L x W):
5.7 x 12.7 cm
  • Varying the 'squeeze' provides controlled delivery
  • One-way inlet valve at the top stops air/dust from being drawn through the tip
  • Nonconductive plastic sleeve-tip prevents scratches and permits cleaning optical lenses, slides, circuit boards,and cuvets
  • Ideal for delicate, difficult-to-clean and inaccessible equipment
  • Environmentally friendly
Specifications
Product TypeDust It™ Blower
Bristle MaterialNone
Bristle TypeNone
Color-codedNo
FormAerosol
Handle TypeNone
ScentUnscented
SterilityNon-sterile
Dimensions (L x W)5.7 x 12.7 cm
For Use With (Application)Powerful bulb duster forcibly blows away dust, lint and dirt with high pressure, pinpoint jets of air
Volume (English)2 oz.
Volume (Metric)56.7 g
Unit SizePK