Carbon Steel Sterile Scalpel Blades
Carbon Steel Sterile Scalpel Blades
Actual product may vary
Fisherbrand
Carbon Steel Sterile Scalpel Blades
Use for convenience in user environments: on the ward, in emergency or trauma units or even out in the field with paramedics. Swann Morton™ Carbon Steel Surgical Scalpel Blades come in single peel packs.
Have Questions?
Catalog NumberSize
FKS11792724# 22
Catalog number FKS11792724
Price (SGD)
88.00
PK
Add to cart
Size:
# 22
Price (SGD)
88.00
PK
Add to cart
  • Carbon steel to BS EN 27740
  • Sterile
Specifications
Blade MaterialCarbon Steel
Product TypeScalpel Blade
Size# 22
For Use With (Equipment)Larger than the # 20 and # 21 the # 22 fits Handles # 4,# 4L, # 4 Graduated & # 6B
SterilitySterile
Certifications/ComplianceCE compliant 93/42/EC
Quantity10/Pk.
Unit SizePK