PTFE Microbore Tubing
PTFE Microbore Tubing
Actual product may vary
Fisherbrand
PTFE Microbore Tubing
Minimize friction and maximize chemical resistance with microbore tubing. Cole-Parmer™ PTFE Microbore Tubing is ideal for extreme temperatures and harsh chemical environments.
Have Questions?
Catalog NumberDiameter (Metric) Inner
FKS117897495.58 mm
Catalog number FKS11789749
Price (SGD)
215.00
EA
Add to cart
Diameter (Metric) Inner:
5.58 mm
Price (SGD)
215.00
EA
Add to cart
PTFE tubing resin is UL-recognized and complies with FDA (21 CFR 177.1550) and USP Class VI requirements.
  • Chemically inert
  • Low permeability
  • Lowest friction coefficient of any tubing
  • Temperature range is -400 to 500°F (-240 to 260°C); melts at 620°F (327°C)
  • Tensile strength is 2500 to 4000 psi
Specifications
Certifications/ComplianceUL-recognized and complies with FDA (21 CFR 177.1550) and USP Class VI requirements.
Diameter (Metric) Inner5.58 mm
MaterialPTFE
Diameter (Metric) Outer1.06 mm
Overall Length (Metric)30.48 m
Thickness (Metric)0.25 mm
For Use With (Application)Use this translucent tubing in extreme temperatures and in harsh chemical environments
Min. Operating Temperature (Metric)-204°C
Packaging Quantity1 Each
ColorTranslucent
Maximum Pressure4000 psi
Max. Operating Temperature (Metric)260°C
Unit SizeEA