Beaker Clamp
Beaker Clamp
Actual product may vary
Fisher Catalogue
Beaker Clamp
Securely holds beakers up to 5L. R&L Enterprises™ Beaker Clamps are manufactured from stainless steel and zinc alloy, with glass fibre fabric covered jaws.
Have Questions?
Catalog NumberFor Use With (Equipment)Length (Metric)
FKS11789027For use with beakers up to 5L360 mm
Catalog number FKS11789027
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
For use with beakers up to 5L
Length (Metric):
360 mm
  • Jaws open to 180mm
  • Rod diameter is 9.5mm
  • Horizontal rod is offset so that large vessels are secured in line with the axis of the vertical support rod
Specifications
Quantity1 Ea.
TypeClamp
MaterialAluminum Alloy, Stainless Steel
Includeswith Glass Fiber Fabric covered jaws
For Use With (Equipment)For use with beakers up to 5L
Length (Metric)360 mm
Jaw TypeSleeves Fiber Heat Resistant Glass
Unit SizeEA