Poly-Net™ HDPE Protective Netting
Poly-Net™ HDPE Protective Netting
Actual product may vary
Fisherbrand
Poly-Net™ HDPE Protective Netting
Prolong the life of all types of glassware with HDPE protective netting. Poly-Net™ HDPE Protective Netting fits snugly over laboratory glassware to reduce breakage and safeguard against flying glass.
Have Questions?
Catalog NumberDiameter (Metric) InnerDescriptionColor
FKS11786038152 to 203 mmClose-Mesh Poly-NetBrown
Catalog number FKS11786038
Price (SGD)
2,570.00
EA
Add to cart
Diameter (Metric) Inner:
152 to 203 mm
Description:
Close-Mesh Poly-Net
Color:
Brown
Price (SGD)
2,570.00
EA
Add to cart
Prolong the life of all types of glassware with HDPE protective netting. Poly-NetTM HDPE Protective Netting fits snugly over laboratory glassware to reduce breakage and safeguard against flying glass.
Use 1/4" to 5/8" diameter sizes for smaller instruments (thermometers, pipettes, electrodes, tubes, syringes, and small bottles). Use 1" to 10" diameter sizes for bottles, flasks, beakers, funnels, desiccators, and columns. Heavy-gauge poly-net is more rigid than close-mesh poly-net; use when extra protection is needed.
  • Tough, resilient HDPE netting
  • Available in rolls of ten
  • Different diameters to fit most vessels
  • Easily cut to needed length
  • Maximum temperature is 125°F (51°C)
Specifications
ColorBrown
Diameter (Metric) Inner152 to 203 mm
Temperature (Metric) Max80°C
For Use With (Application)Extra Protection
Model0436-BROWN
Packaging Quantity1 Each
DescriptionClose-Mesh Poly-Net
For Use WithFor Bottles, Flasks, Beakers, Funnels, Desiccators, and Columns
Overall Length (Metric)50 m
MaterialHDPE
Unit SizeEA