Stainless Steel Freezer Rack
Stainless Steel Freezer Rack
Actual product may vary
Fisherbrand
Stainless Steel Freezer Rack
Store assay plates and PCR racks securely in steel freezer racks. Stainless Steel Freezer Racks are available in two styles: locking rod racks that allow stacking of assay plates or horizontal drawer racks for easy box access. Both styles provide 16 compartments of secure cryostorage.
Have Questions?
Catalog NumberShelves
FKS11786013Pivoting Pull-out Drawers
Catalog number FKS11786013
Price (SGD)
1,433.00
EA
Add to cart
Shelves:
Pivoting Pull-out Drawers
Price (SGD)
1,433.00
EA
Add to cart
For cryostorage in upright freezers.
  • Constructed of corrosion-resistant stainless steel.
  • For Assay Plate, PCR rack and cryogenic tube box storage.
  • Designed to fit in most standard upright freezers.
  • Sturdy containers with 4 levels of storage, providing 16 total storage compartments.
  • Wire handles on the front allow for easy removal of the rack from the freezer.
Locking Rod Freezer Rack:
  • Rack includes four vertical locking rods over the side opening to secure contents in place.
  • Allows stacking of short assay plates in each slot (not included):
    • 96-Well Assay plates (holds 3 per compartment, up to 48 total)
    • 24-Well Assay plates (holds 2 per compartment, up to 32 total)
    • 96-Deep Well Assay plates (holds 1 per compartment, up to 16 total)
    • 96-Well PCR racks (holds 1 per compartment, up to 16 total)
Horizontal Drawer Freezer Rack
  • Horizontal drawer system allow for easy box access
  • Drawers feature a stop that pivots down when fully extended
  • Polycarbonate drawer handles provide a user-friendly grip
  • Includes a slot and label for drawer/sample identification
  • Ideal for standard boxes up to 133 x 133 x 53 mm in size
Specifications
Product TypeFreezer Rack
Array4 x 4
AutoclavableNot Autoclavable
ColorSilver
HoldsBoxes (2 in.)
MaterialStainless Steel
Positions16
ShapeRectangular
ShelvesPivoting Pull-out Drawers
StackableYes
SterilityNon-sterile
For Use With (Application)For standard PCR rack and cryogenic rack (up to 133 x 133 x 53 mm) storage in upright freezers
No. of Racks4
No. per Pack1 Each
Quantity1 Ea.
Dimensions (L x W x H)571 x 139 x 238 mm
Length (Metric)571 mm
Length (English)22.5 in.
Width (Metric)139 mm
Width (English)5.5 in.
Height (Metric)238 mm
Height (English)9.4 in.
Unit SizeEA