Check Order Status
HDPE Stackable Pail
Actual product may vary
Fisherbrand
HDPE Stackable Pail
Have Questions?
Catalog Number
Capacity (Metric)
FKS11785328
16.1 L
Catalog number
FKS11785328
Price (SGD)
-
Capacity (Metric):
16.1 L
Specifications
Product Type
Stackable Pail
Capacity (Metric)
16.1 L
Material
HDPE
Certifications/Compliance
FDA Certified
Height (Metric)
368.3 mm
Height (Metric) With Closure
368.3 mm
Quantity
1 Ea.
Width (Metric)
247.6 mm