L/S™ Platinum-Cured Silicone Pump Tubing, spooled
L/S™ Platinum-Cured Silicone Pump Tubing, spooled
Actual product may vary
Fisherbrand
L/S™ Platinum-Cured Silicone Pump Tubing, spooled
Reduce scrap with spooled tubing. Masterflex™ L/S™ Platinum-Cured Silicone Pump Tubing features a smooth surface and low protein binding levels, ideal for pharmaceutical, biotechnology, peristaltic pumping and food applications.
Have Questions?
Catalog NumberTubing Size
FKS1178517824
Catalog number FKS11785178
Price (SGD)
10,217.00
EA
Add to cart
Tubing Size:
24
Price (SGD)
10,217.00
EA
Add to cart
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
  • Ensure that you always have enough tubing on hand
  • Slightly greater clarity
  • Preferred by pharmaceutical and biotechnology industries
  • Fewer potential leachables
  • USP Class VI
  • FDA compliant
Specifications
Diameter (Metric) Inner6.4 mm
MaterialPlatinum-cured Silicone
For Use With (Application)Pharmaceutical and biotechnology industries
Min. Operating Temperature (Metric)-50°C
Packaging Quantity1 Ea.
Tubing Size24
Flow Rate2.8 to 1700 mL/min.
Hose Barb1/4 in.
Maximum Pressure40 psi
Vacuum Level660 mm Hg
Max. Operating Temperature (Metric)230°C
Product TypePump Tubing
Unit SizeEA