Interlocking Cube Rack
Interlocking Cube Rack
Actual product may vary
Fisherbrand
Interlocking Cube Rack
Work with different tube sizes in the same plane. Fisherbrand™ Interlocking Cube Racks accommodate different tube sizes on each side. Connect cubes together, oriented to any side, for a customized, user-friendly rack. Constructed of strong, autoclavable polypropylene in five assorted colors.
Have Questions?
Catalog NumberColor
FKS11784726Assorted
Catalog number FKS11784726
Price (SGD)
200.00
PK
Add to cart
Color:
Assorted
Price (SGD)
200.00
PK
Add to cart
  • Enables different sized tubes to be accessible all in the same plane
  • User friendly cubes connect together for a customizable rack size
  • Simply rotate the cube to access the needed tube well array
    • 4 x 50 mL tubes
    • 10 x 15 mL tubes
    • 12 x 12 mL tubes
    • 16 x 1.5/2.0 mL microtubes
  • Constructed of heavy duty polypropylene in five assorted colors
  • Autoclavable
Specifications
Product TypeMicrotube Rack
Closure IncludedNo
DescriptionInterlocking Tube Racks, 4-Sided
AutoclavableAutoclavable
ColorAssorted
HoldsTubes (4 x 50 mL or 10 x 15 mL or 12 x 12 mL or 16 x 1.5/2.0 mL Microtubes)
MaterialPolypropylene
ShapeCubical
For Use With (Equipment)Microcentrifuge Tube
No. per Pack5 Pack
Quantity5 Pack
Length (Metric)108 mm
Width (Metric)113 mm
Height (Metric)113 mm
Unit SizePK