Check Order Status
Rubber Small Volume Pacifier
Actual product may vary
Fisherbrand
Rubber Small Volume Pacifier
Use this small volume pacifier for a variety of applications. Saint Gobain Performance Plastics™ Rubber Small Volume Pacifier are compatible with a wide range of pipettes.
Have Questions?
Catalog Number
Color
Capacity (Metric)
FKS11784498
White
2 mL
Catalog number
FKS11784498
Price (SGD)
-
Color:
White
Capacity (Metric):
2 mL
For liquid handling
Specifications
Quantity
10/Pk.
Type
Small Volume Pacifier
Material
Rubber
Color
White
Capacity (Metric)
2 mL